Learn More
Leica Biosystems 1ml NCL-L-CD20-L26
Supplier:  Leica Biosystems CD20L26LCE
Encompass
Antigen Background The CD20 antigen is a non-glycosylated phosphoprotein of approximately 33kD which is expressed on normal and malignant human B cells and is thought to act as a receptor during B cell activation and differentiation. CD20 antigen has been reported to be expressed on normal B cells from peripheral blood, lymph node, spleen, tonsil, bone marrow, acute leukemias and chronic lymphocytic leukemias. Disclaimer CD20 is recommended for the detection of specific antigens of interest in normal and neoplastic tissues, as an adjunct to conventional histopathology using non-immunologic histochemical stains.
Catalog No. 50-255-2325