Learn More
Leica Biosystems 1ml NCL-L-CD4-368
Supplier:  Leica Biosystems CD4368LU
Encompass
Antigen BackgroundThe CD4 molecule (T4) is a single chain transmembrane glycoprotein with a molecular weight of 59 kD. The CD4 antigen is expressed on a T cell subset (helper/inducer) representing 45% of peripheral blood lymphocytes and at a lower level on monocytes and germinal center macrophages. Most cases of cutaneous T cell lymphoma, including mycosis fungoides, express the CD4 antigen and HTLV-1 associated adult T cell leukemia/lymphoma is also generally CD4 positive.DisclaimerCD4 is recommended for the detection of specific antigens of interest in normal and neoplastic tissues, as an adjunct to conventional histopathology using non-immunologic histochemical stains.
Catalog No. 50-255-2311