Learn More
Leica Biosystems 1ml NCL-L-CK5/6/8/18
Supplier:  Leica Biosystems CK5/6/8/18L
Encompass
Antigen Background NCL-CK5/6/8/18, NCL-L-CK5/6/8/18 and RTU-CK5/6/8/18 react with human cytokeratins 5, 6, 8 and 18.These products are cocktails of monoclonal antibodies designed to recognize cytokeratins reported to be expressed in almost all epithelial tissues. Disclaimer Cytokeratin, Multi (5/6/8/18) is recommended for the detection of specific antigens of interest in normal and neoplastic tissues, as an adjunct to conventional histopathology using non-immunologic histochemical stains.
Catalog No. 50-255-2287