Learn More
Leica Biosystems 1ml NCL-L-DOG-1
Supplier:  Leica Biosystems DOG1LCE
Encompass
Antigen Background DOG-1, a 986 amino acid protein of unknown function, is expressed predominantly on the plasma membrane of gastrointestinal stromal tumors (GISTs) and is rarely expressed in other soft tissue tumors, which, due to appearance, can be confused with GISTs. Reactivity for DOG-1 has been suggested to aid in the identification of GISTs, including Platelet-Derived Growth Factor Receptor Alpha mutants that fail to express KIT antigen. Product Specific Information The use of PBS-based diluents may result in increased background staining. Disclaimer DOG-1 is recommended for the detection of specific antigens of interest in normal and neoplastic tissues, as an adjunct to conventional histopathology using non-immunologic histochemical stains.
Catalog No. 50-255-2277