Learn More
Leica Biosystems 1ml NCL-L-EGFR
Supplier:  Leica Biosystems EGFRLCE
Encompass
Antigen BackgroundEpidermal growth factor receptor (EGFR) is a transmembrane protein receptor of 170 kD with tyrosine kinase activity. Increased levels of EGFR are reported to be linked with malignant transformation of squamous cells eg in squamous cell carcinoma of the lung, head, neck, skin, cervix and esophagus. EGFR may also play a role in the development and progression of hepatocellular carcinomas where recurrence rates are higher in EGFR-positive cases. This correlation has similarly been reported in colorectal cancers where EGFR, produced by tumor cells, plays an important role in the invasiveness and proliferation of colorectal cancers. The majority of published studies of EGFR expression in human breast cancer has similarly shown an association with EGFR expression where it is inversely related to estrogen receptor status.Product Specific InformationClone EGFR.113 is raised to the extracellular domain of the EGFR molecule.DisclaimerEpidermal Growth Factor Receptor is r
Catalog No. 50-255-2262