Learn More
Leica Biosystems 1ml NCL-L-ER-6F11 Class II
Supplier:  Leica Biosystems ER6F11LF
Encompass
Antigen BackgroundEstrogen receptor (ER) content of breast cancer tissue is an important parameter in the prediction of prognosis and response to endocrine therapy. The introduction of highly specific monoclonal antibodies to ER has allowed the determination of receptor status of breast tumors to be carried out in routine histopathology laboratories.Product Specific InformationClone 6F11 is raised to the full length alpha form of the estrogen receptor molecule.DisclaimerEstrogen Receptor is recommended for the detection of specific antigens of interest in normal and neoplastic tissues, as an adjunct to conventional histopathology using non-immunologic histochemical stains.
Catalog No. 50-255-2252