Learn More
Leica Biosystems 1ml NCL-L-VIM-572
Supplier:  Leica Biosystems VIM572LCE
Encompass
Antigen Background Eukaryotic cells contain a number of types of cytoplasmic filamentous proteins, microtubule, microfilaments and intermediate-sized filaments (IF). Vimentin, a 57 kD protein that is an intermediate filament is reported to be expressed in most cells of mesenchymal origin, including fibroblasts, endothelial cells, smooth muscle, melanocytes as well as T and B lymphocytes. Disclaimer Vimentin is recommended for the detection of specific antigens of interest in normal and neoplastic tissues, as an adjunct to conventional histopathology using non-immunologic histochemical stains.
Catalog No. 50-255-1947