Learn More
USA Scientific Inc .1mL PCR 8tb white strip
Catalog No. 502745543
Encompass
0.1 mL TempAssure PCR low profile flex-free 8-tube strips with individually attached, optically clear flat caps. 120 strips of tubes with caps per pack.
Catalog No. 50-274-5543 Supplier USA Scientific Inc Supplier No. 14022380
0.1 mL TempAssure PCR low profile flex-free 8-tube strips with individually attached, optically clear flat caps. 120 strips of tubes with caps per pack.