Learn More
CELLTREAT Scientific Products 1mL Pipet, Open End, Individually Wrapped, Bag, Sterile
SDP
Catalog No. NC3433441
Encompass_Preferred
"Ideal for viscous and high particulate solutionsWide, open end is ideal for slurries and gelsAll serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934; Accuracy is ±2%Bi-directional (ascending & descending) graduations are printed in bold easy-to-read black inkPremium polymer fiber barrier plug outperforms cotton plugsColor coded stripes and packaging enable quick volume identificationPrinted indicator arrow for quickly locating negative graduations when using a pipet controller"
Catalog No. NC3433441 Supplier CELLTREAT Scientific Products Supplier No. 229221B
"Ideal for viscous and high particulate solutionsWide, open end is ideal for slurries and gelsAll serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934; Accuracy is ±2%Bi-directional (ascending & descending) graduations are printed in bold easy-to-read black inkPremium polymer fiber barrier plug outperforms cotton plugsColor coded stripes and packaging enable quick volume identificationPrinted indicator arrow for quickly locating negative graduations when using a pipet controller"