Learn More
CELLTREAT Scientific Products 1mL Serological Pipet, Individual Plastic/Plastic Wrapper Packed in Bags, Sterile pipette
SDP
Supplier:  CELLTREAT Scientific Products 229201A
Encompass_Preferred
CELLTREAT Scientific Products 1mL Serological Pipet. Peel-to-open plastic/plastic wrappers are the ideal option for eliminating particulate; proven benefit over traditional paper/plastic wrapper. Wrappers reduce static cling versus other brands of plastic wrap pipets. All plastic wrapper can be wiped with alcohol without smearing ink or penetrating paper. All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934. Bi-directional (positive and negative) graduations are printed in bold easy-to-read ink. Premium material barrier plug. Color coded stripes and packaging enable quick volume identification. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, individually wrapped 100/bag, 800/case.
Catalog No. 50-828-754