Learn More
Waters Corp Waters Corporation Quick Load Replacement Inserts, 1 mL, 96-Well Plate, Glass
Catalog No. 50780696 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These quick load, 1 mL glass inserts are designed for use with 96-well plates, offering a precise solution for applications with limited sample volume. They are strictly QC tested for cleanliness and dimensional tolerances, ensuring functionality and preventing contamination. The inserts effectively accommodate small sample volumes and help prevent compounds from sticking to plastic surfaces.
  • Quick load, 1 mL glass inserts for 96-well plates.
  • Ideal for applications with limited sample volume.
  • Enable fast application to well plates.
  • QC tested for cleanliness and dimensional precision.
  • Helps prevent compounds from adhering to plastic surfaces.
Catalog No. 50-780-696 Supplier Waters Corp Supplier No. 186001436
These quick load, 1 mL glass inserts are designed for use with 96-well plates, offering a precise solution for applications with limited sample volume. They are strictly QC tested for cleanliness and dimensional tolerances, ensuring functionality and preventing contamination. The inserts effectively accommodate small sample volumes and help prevent compounds from sticking to plastic surfaces.
  • Quick load, 1 mL glass inserts for 96-well plates.
  • Ideal for applications with limited sample volume.
  • Enable fast application to well plates.
  • QC tested for cleanliness and dimensional precision.
  • Helps prevent compounds from adhering to plastic surfaces.