Learn More
MyBioSource Inhibin beta E (INHBE) Polyclonal Antibody
SDP
Supplier:  MyBioSource MBS20034661 ML
Encompass_Preferred
MyBioSource offers a rabbit polyclonal antibody against human inhibin beta E (INHBE) for research use. This antibody detects endogenous levels of total INHBE protein. INHBE is a member of the TGF-beta superfamily and is involved in follicle development. This antibody can be used for ELISA, WB, IHC, and ICC/IF applications. It is provided at 100 μg in PBS with 0.02% sodium azide and 0.5% BSA.
Catalog No. NC2753531