Learn More
Bellco Glass, Inc. 1mL Sero Pipet Individ Wrapped
SDP
Catalog No. 502329956
Encompass_Preferred
All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets.Specifications Bidirectional positive and negative graduations are printed in bold black ink10mL wide tip Serological pipet for viscous solutions. Wide tip is 4x larger than standard 10mL pipet 25 amp 50mL pipet features an antidrip tip for accurate delivery Color Coded stripes and packing enable quick volume identification Printed indicator arrow for quickly locating negative graduations when using a pipet controller. 0.3mL graduations 100 individually wrapped bag pack Case Quantity 500nbsp
Catalog No. 50-232-9956 Supplier Bellco Glass, Inc. Supplier No. 229001B00
All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets.Specifications Bidirectional positive and negative graduations are printed in bold black ink10mL wide tip Serological pipet for viscous solutions. Wide tip is 4x larger than standard 10mL pipet 25 amp 50mL pipet features an antidrip tip for accurate delivery Color Coded stripes and packing enable quick volume identification Printed indicator arrow for quickly locating negative graduations when using a pipet controller. 0.3mL graduations 100 individually wrapped bag pack Case Quantity 500nbsp