Learn More
CROSSTEX INTERNATIONAL 1mL Spore Ampules, 50/box, Population:10^4
Catalog No. NC1619314
Encompass_Preferred
Crosstex 1 mL Spore Ampoules are Self-Contained Biological Indicators for monitoring steam processes consisting of a hermetically sealed glass ampoule which contains modified Trypticase Soy Broth (TSB) with a pH indicator and Geobacillus stearothermophilus spores.
Catalog No. NC1619314 Supplier CROSSTEX INTERNATIONAL Supplier No. NMSA15004
Crosstex 1 mL Spore Ampoules are Self-Contained Biological Indicators for monitoring steam processes consisting of a hermetically sealed glass ampoule which contains modified Trypticase Soy Broth (TSB) with a pH indicator and Geobacillus stearothermophilus spores.