Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One 1mL Sterile Serological Pipets
GREENER_CHOICE

Catalog No. 07000362 Shop All Greiner Bio-One Products
Change view
Click to view available options
Packaging Type:
Bulk
Individual Paper/Plastic Wrap
Individual Plastic Wrap
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Type
07-000-362 Individual Paper/Plastic Wrap
07-000-365 Bulk
07-000-364 Individual Plastic Wrap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 07-000-362 Supplier Greiner Bio-One Supplier No. 604181
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Drip-free pipetting design with high optical clarity and maximum accuracy.

  • High-grade polystyrene
  • Clear, easily legible graduation
  • Colored vertical stripe for ease in reading the volume
  • With synthetic fibrous filter material for protection against
TRUSTED_SUSTAINABILITY

Specifications

Aspirator Plug Yes
Description Shape: Standard
Material Polystyrene
Sterility Sterile
Graduated Yes
Graduations 1/100
Includes Synthetic Fibrous Filter Material
Negative Graduations Yes
Quantity 1000 Pk.
Packaging Type Individual Paper/Plastic Wrap
Color Clear
Volume (Metric) 1 mL
Type Serological Pipet
Disposable Single-use
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.