Learn More
Trajan Scientific L-MARK Brand 1.0mL Syringe, RN, GT, Pt.3, PTFE Plunger Tip, 19g, 4"" Needle
Catalog No. 50635271
Encompass_Preferred
L-MARK Brand Syringe, 1mL, 4- inch Needle, 19 Gauge, Point Style 3, Removable Needle, (Single Needle in kit), can use LMK.2831028 Needles 25ul-1.0ml Made in Germany.
Catalog No. 50-635-271 Supplier Trajan Scientific Supplier No. LMK.2620014
L-MARK Brand Syringe, 1mL, 4- inch Needle, 19 Gauge, Point Style 3, Removable Needle, (Single Needle in kit), can use LMK.2831028 Needles 25ul-1.0ml Made in Germany.