Learn More
Analytical Sales & Services 1mL Tapered/Flat-Bottom Clear Glass Inserts in Vial Loader
Catalog No. 5000806210
Encompass
1mL Clear Glass Tapered, Flat-Bottom Inserts in Vial Loader\n\n• Vial loader contains 96 inserts\n\nQty/Pkg: Each
Catalog No. 50-008-06210 Supplier Analytical Sales & Services Supplier No. 96405VL
1mL Clear Glass Tapered, Flat-Bottom Inserts in Vial Loader\n\n• Vial loader contains 96 inserts\n\nQty/Pkg: Each