Learn More
Analytical Sales & Services 1ml TrueTaper 96-Well Collection Plate with 50uL Tapered-Bottom Wells, Irradiated
Catalog No. 5000806236
Encompass
1mL TrueTaper 96-well Collection Plate, Irradiated\n\n• Unique Tapered Bottom for Maximum Sample Recovery\n• Round Wells with 50µL TAPERED Bottoms (1mL Total Volume per Well)\n• Polypropylene\n• Irradiated\n• Made in USA of domestically sourced and certified materials\n\nQty/Pkg: 25\n\n 
Catalog No. 50-008-06236 Supplier Analytical Sales & Services Supplier No. 968810RAD
1mL TrueTaper 96-well Collection Plate, Irradiated\n\n• Unique Tapered Bottom for Maximum Sample Recovery\n• Round Wells with 50µL TAPERED Bottoms (1mL Total Volume per Well)\n• Polypropylene\n• Irradiated\n• Made in USA of domestically sourced and certified materials\n\nQty/Pkg: 25\n\n 