Learn More
Analytical Sales & Services 1mL TrueTaper 96-Well Collection Plate with 50uL Tapered-Bottom Wells, with SiliGuard™ Low Bind Coating, Irradiated
Catalog No. 5000806252
Encompass
1mL TrueTaper 96-well Collection Plate with SiliGuard­™, Irradiated\n\n• Unique Tapered Bottom for Maximum Sample Recovery\n• Round Wells with 50µL TAPERED Bottoms (1mL Total Volume per Well)\n• Polypropylene\n• SiliGuard­™ Low Bind Coating\n• Irradiated\n• Made in USA of domestically sourced and certified materials\n\nQty/Pkg: 25
Catalog No. 50-008-06252 Supplier Analytical Sales & Services Supplier No. 968810GCRAD
1mL TrueTaper 96-well Collection Plate with SiliGuard­™, Irradiated\n\n• Unique Tapered Bottom for Maximum Sample Recovery\n• Round Wells with 50µL TAPERED Bottoms (1mL Total Volume per Well)\n• Polypropylene\n• SiliGuard­™ Low Bind Coating\n• Irradiated\n• Made in USA of domestically sourced and certified materials\n\nQty/Pkg: 25