Learn More
Ace Glass, Inc. 1ML VOLUMETRIC BULB 1ML VOLUMETRIC BULB
SDP
Catalog No. 50975868
Encompass_Preferred
1mL volumetric bulb for use with ACE Automatic Dispensing Pipet. Bulbs have a short tipping angle and give reproduceable volumes.
Catalog No. 50-975-868 Supplier Ace Glass, Inc. Supplier No. 800415
1mL volumetric bulb for use with ACE Automatic Dispensing Pipet. Bulbs have a short tipping angle and give reproduceable volumes.