Learn More
Ace Glass, Inc. Adapter, glass capillary, adjustable electrode, complete, nylon bushing with 1.0mm hole
SDP
Catalog No. 50971665
Encompass_Preferred
Ace-Thred Glass Capillary Adjustable Electrode Adapter, sold complete with nylon bushing with 1.0mm hole and silicone rubber three-layer septum. Adapter is for use with platinum or other electrodes that normally would be step-sealed to glass. The use of a threaded bushing and a silicone rubber septum for sealing allow for the electrode to be adjusted for depth or removed, if desired. Capillary of glass is 11/2-2mm with 76mm of stem for sealing to apparatus. Overall length of glass is approximately 140mm.
Catalog No. 50-971-665 Supplier Ace Glass, Inc. Supplier No. 503720
Add to Cart
Add to Cart
Ace-Thred Glass Capillary Adjustable Electrode Adapter, sold complete with nylon bushing with 1.0mm hole and silicone rubber three-layer septum. Adapter is for use with platinum or other electrodes that normally would be step-sealed to glass. The use of a threaded bushing and a silicone rubber septum for sealing allow for the electrode to be adjusted for depth or removed, if desired. Capillary of glass is 11/2-2mm with 76mm of stem for sealing to apparatus. Overall length of glass is approximately 140mm.