Learn More
Alkali Scientific 1mm Electroporation Cuvettes, Round Lid, Sterile, 50/cs
SDP
Catalog No. 502389536
Encompass_Preferred
Manufactured from high quality durable polycarbonate material which has been tested to optimize transfection efficiencies. Precision built to give exacting tolerances ensuring reproducible results. Polished aluminum plates reduce static and arcing for optimum performance. Individually wrapped and gamma irradiated providing guaranteed sterility. Compatible with most Electroporation devices. Individually color coded cuvette caps identify gap widths of 1mm. Each bag contains 50 individually wrapped sterile cuvettes.
Catalog No. 50-238-9536 Supplier Alkali Scientific Supplier No. EP101001
Manufactured from high quality durable polycarbonate material which has been tested to optimize transfection efficiencies. Precision built to give exacting tolerances ensuring reproducible results. Polished aluminum plates reduce static and arcing for optimum performance. Individually wrapped and gamma irradiated providing guaranteed sterility. Compatible with most Electroporation devices. Individually color coded cuvette caps identify gap widths of 1mm. Each bag contains 50 individually wrapped sterile cuvettes.