Learn More
USA Scientific Inc Electroporation cuvette, 0.1 cm gap
Catalog No. NC9763528
Encompass_Preferred
Electroporation cuvette, 0.1 cm gap width, white cap, individually wrapped, sterile, 50/bag
Catalog No. NC9763528 Supplier USA Scientific Inc Supplier No. 91041050
Electroporation cuvette, 0.1 cm gap width, white cap, individually wrapped, sterile, 50/bag