Learn More
BIOSMITH Consumables. 1mm gap electroporation cuvette. Fits all commercial electroporators. Single packaged. 50 per pack
Catalog No. NC2980199
Encompass
Consumables. 1mm gap electroporation cuvette. Fits all commercial electroporators. Single packaged. 50 per pack
Catalog No. NC2980199 Supplier BIOSMITH Supplier No. 72001
Consumables. 1mm gap electroporation cuvette. Fits all commercial electroporators. Single packaged. 50 per pack