Learn More
Ace Glass, Inc. 1MM MICRO STOPCOCK (GB)
SDP
Catalog No. 50976123
Encompass_Preferred
1mm micro stopcock for micro chemical apparatus. Solid glass plug and barrel are lapped together providing a precision fit. Plugs are interchangeable within the meaning of ASTM Standard E 675.
Catalog No. 50-976-123 Supplier Ace Glass, Inc. Supplier No. 813910
1mm micro stopcock for micro chemical apparatus. Solid glass plug and barrel are lapped together providing a precision fit. Plugs are interchangeable within the meaning of ASTM Standard E 675.