Learn More
Azzota Corp 1mm Pathlength Std Cuvette Q
Catalog No. 5000887090
Encompass_Preferred
1mm Pathlength Standard Cuvettes 0 35ml Quartz (190nm-2500nm) with Teflon stopper but round bottom
Catalog No. 50-008-87090 Supplier Azzota Corp Supplier No. Q111
1mm Pathlength Standard Cuvettes 0 35ml Quartz (190nm-2500nm) with Teflon stopper but round bottom