Learn More
Chemglass Life Sciences Stopcock, Straight Bore, PTFE Plug, 1mm Bore, 11/25 Plug Size, 7mm OD Sidearms
SDP
Catalog No. 501531732
Encompass_Preferred
Stopcock, Straight Bore, PTFE Plug, 1mm Bore, 11/25 Plug Size, 7mm OD Sidearms. Straight bore stopcock having a 1:5 taper solid PTFE plug.
Catalog No. 50-153-1732 Supplier Chemglass Life Sciences Supplier No. CG42401
Stopcock, Straight Bore, PTFE Plug, 1mm Bore, 11/25 Plug Size, 7mm OD Sidearms. Straight bore stopcock having a 1:5 taper solid PTFE plug.