Learn More
Azzota Corp 1mm Pathlength Standard Cuvettes - 0.35Ml Pathlength - 1mm Inside Width - 10mm Volume - 0.35Ml Dimension - 45X12.5X3.5mm With Cover And Round Bottom Quartz (190Nm-2500Nm)
Catalog No. 507532892
Encompass_Preferred
1mm Pathlength Standard Cuvettes - 0.35ml Pathlength - 1mm Inside width - 10mm Volume - 0.35ml Dimension - 45x12.5x3.5mm With cover and round bottom Quartz (190nm-2500nm)
Catalog No. 50-753-2892 Supplier Azzota Corp Supplier No. Q101
1mm Pathlength Standard Cuvettes - 0.35ml Pathlength - 1mm Inside width - 10mm Volume - 0.35ml Dimension - 45x12.5x3.5mm With cover and round bottom Quartz (190nm-2500nm)