Learn More
Teknova 1mM Sodium Citrate. pH 6.4, 1000mL. RNase and Endotoxin Tested. Sterile. 2 pack.
SDP
Catalog No. 502387775
Encompass
Sodium citrate solution is a commonly used reagent in life science. It has many uses, such as making media for the growth of microorganisms, lysing cells, and precipitating nucleic acids.
Catalog No. 50-238-7775 Supplier Teknova Supplier No. S85132PK
Sodium citrate solution is a commonly used reagent in life science. It has many uses, such as making media for the growth of microorganisms, lysing cells, and precipitating nucleic acids.