Learn More
Alkali Scientific 1mm Electroporation Cuvettes, Square Lid, Sterile, 50/cs
SDP
Catalog No. 502388945
Encompass_Preferred
Manufactured from high quality durable polycarbonate material which has been tested to optimize transfection efficiencies. Precision built to give exacting tolerances ensuring reproducible results. Polished aluminum plates reduce static and arcing for optimum performance. Individually wrapped and gamma irradiated providing guaranteed sterility. Compatible with most Electroporation devices. Individually color coded cuvette caps identify gap widths of 1mm. Each bag contains 50 individually wrapped sterile cuvettes.
Catalog No. 50-238-8945 Supplier Alkali Scientific Supplier No. EP401001
Manufactured from high quality durable polycarbonate material which has been tested to optimize transfection efficiencies. Precision built to give exacting tolerances ensuring reproducible results. Polished aluminum plates reduce static and arcing for optimum performance. Individually wrapped and gamma irradiated providing guaranteed sterility. Compatible with most Electroporation devices. Individually color coded cuvette caps identify gap widths of 1mm. Each bag contains 50 individually wrapped sterile cuvettes.