Learn More
Ace Glass, Inc. 1MM STOPCOCK GB 1MM STOPCOCK GB
SDP
Catalog No. 50976113
Encompass_Preferred
Glass 1mm straight bore stopcock with a solid glass plug and a tapered flange, which insures maximum strength at the seal. The bore of the side arms is equal to or slightly larger than the bore of the plug.
Catalog No. 50-976-113 Supplier Ace Glass, Inc. Supplier No. 813702
Glass 1mm straight bore stopcock with a solid glass plug and a tapered flange, which insures maximum strength at the seal. The bore of the side arms is equal to or slightly larger than the bore of the plug.