Learn More
Chemglass Life Sciences Stopcock, Straight Bore, Micro, PTFE Plug, 1mm Bore, 10/15 Plug Size, 6mm OD Sidearms
SDP
Catalog No. CG42501
Encompass_Preferred
Stopcock, Straight Bore, Micro, PTFE Plug, 1mm Bore, 10/15 Plug Size, 6mm OD Sidearms. Used in the fabrication of small scale apparatus, especially apparatus that has weight restrictions. PTFE plug has a 1:5 taper.
Catalog No. CG42501 Supplier Chemglass Life Sciences Supplier No. CG42501
Stopcock, Straight Bore, Micro, PTFE Plug, 1mm Bore, 10/15 Plug Size, 6mm OD Sidearms. Used in the fabrication of small scale apparatus, especially apparatus that has weight restrictions. PTFE plug has a 1:5 taper.