Learn More
Cayman Chemical 1-Methyl-1,4-dihydronicotinamide, Purity: ≥90%, CAS: 17750-23-1, 50 mg
SDP
Catalog No. 502993791
Encompass_Preferred
A nicotinamide derivative; inhibits β-elimination activity of hSR (IC50 = 177 μM)
Catalog No. 50-299-3791 Supplier Cayman Chemical Supplier No. 2322650 MG
A nicotinamide derivative; inhibits β-elimination activity of hSR (IC50 = 177 μM)