Learn More
Cayman Chemical 1MyrIstoyl2hydroxysnglyc 100mg
SDP
Catalog No. 502936799
Encompass_Preferred
A lysophospholipid; has been used in the generation of micelles, liposomes, and other types of artificial membranes, including lipid-based drug carrier systems
Catalog No. 50-293-6799 Supplier Cayman Chemical Supplier No. 25587100 MG
A lysophospholipid; has been used in the generation of micelles, liposomes, and other types of artificial membranes, including lipid-based drug carrier systems