Learn More
Cayman Chemical 1-Myristoyl-2-hydroxy-sn-glycero-3-PC, Purity: ≥95%, CAS: 20559-16-4, 10 mg
SDP
Catalog No. 502983280
Encompass_Preferred
A saturated 14:0 lysophosphatidylcholine that consists of one chain of myristic acid at the sn-1 position
Catalog No. 50-298-3280 Supplier Cayman Chemical Supplier No. 2095810 MG
A saturated 14:0 lysophosphatidylcholine that consists of one chain of myristic acid at the sn-1 position