Learn More
Cayman Chemical 1-Myristoyl-2-hydroxy-sn-glycero-3-PG (sodium salt), Purity: ≥95% (9:1 mixture of 1-acyl and 2-acyl), CAS: 326495-21-0, 1 g
SDP
Catalog No. 502935778
Encompass_Preferred
A lysophospholipid; has been used in the generation of micelles, liposomes, and other types of artificial membranes, including lipid-based drug carrier systems
Catalog No. 50-293-5778 Supplier Cayman Chemical Supplier No. 255871 G
A lysophospholipid; has been used in the generation of micelles, liposomes, and other types of artificial membranes, including lipid-based drug carrier systems