Learn More
Cayman Chemical 1-Myristoyl-2-hydroxy-sn-glycero-3-PE, Purity: ≥98%, CAS: 123060-40-2, 5 mg
SDP
Catalog No. 502936079
Encompass_Preferred
A naturally occurring lysophospholipid; induces transient increases in intracellular calcium in PC12 cells; serum levels are elevated in patients with malignant breast cancer compared to healthy controls
Catalog No. 50-293-6079 Supplier Cayman Chemical Supplier No. 255915 MG
A naturally occurring lysophospholipid; induces transient increases in intracellular calcium in PC12 cells; serum levels are elevated in patients with malignant breast cancer compared to healthy controls