Learn More
Cayman Chemical 1MyrIstoyl2Oleoyl3Docosa 10mg
SDP
Catalog No. 502938012
Encompass_Preferred
A triacylglycerol; has been used as a reagent to study the regioselectivity of thermal chlorination reactions between triacylglycerols and organochlorines that result in the formation of MCPD diesters
Catalog No. 50-293-8012 Supplier Cayman Chemical Supplier No. 2738210 MG
A triacylglycerol; has been used as a reagent to study the regioselectivity of thermal chlorination reactions between triacylglycerols and organochlorines that result in the formation of MCPD diesters