Learn More
Cayman Chemical 1-Myristoyl-2-Oleoyl-3-Palmitoyl-rac-glycerol, Purity: ≥95%, CAS: 99131-43-8, 50 mg
SDP
Catalog No. 502938664
Encompass_Preferred
A triacylglycerol; has been found in butterfat and the fat body of male B. hortorum bumblebees; has also been found as a minor triacylglycerol preserved in the subcutaneous fat of the 5,300-year-old Iceman mummy
Catalog No. 50-293-8664 Supplier Cayman Chemical Supplier No. 2923350 MG
A triacylglycerol; has been found in butterfat and the fat body of male B. hortorum bumblebees; has also been found as a minor triacylglycerol preserved in the subcutaneous fat of the 5,300-year-old Iceman mummy