Learn More
Cayman Chemical 1MyrIstoyl2PalmItoyl3But 10mg
SDP
Catalog No. 502935575
Encompass_Preferred
A triacylglycerol containing myristic, palmitic, and butyric acids at the sn-1, sn-2, and sn-3 positions, respectively; has been detected in human breast milk
Catalog No. 50-293-5575 Supplier Cayman Chemical Supplier No. 2682110 MG
A triacylglycerol containing myristic, palmitic, and butyric acids at the sn-1, sn-2, and sn-3 positions, respectively; has been detected in human breast milk