Learn More
Cayman Chemical 1-Myristoyl-2-Palmitoyl-3-Butyryl-rac-glycerol, Purity: ≥98%, CAS: 1122578-89-5, 5 mg
SDP
Catalog No. 502932951
Encompass_Preferred
A triacylglycerol containing myristic, palmitic, and butyric acids at the sn-1, sn-2, and sn-3 positions, respectively; has been detected in human breast milk
Catalog No. 50-293-2951 Supplier Cayman Chemical Supplier No. 268215 MG
A triacylglycerol containing myristic, palmitic, and butyric acids at the sn-1, sn-2, and sn-3 positions, respectively; has been detected in human breast milk