Learn More
Cayman Chemical 1-Myristoyl-3-Butyryl-rac-glycerol, Purity: ≥98%, CAS: 100732-08-9, 50 mg
SDP
Catalog No. 502932968
Encompass_Preferred
A diacylglycerol containing myristic acid at the sn-1 position and butyric acid at the sn-3 position
Catalog No. 50-293-2968 Supplier Cayman Chemical Supplier No. 2679450 MG
A diacylglycerol containing myristic acid at the sn-1 position and butyric acid at the sn-3 position