Learn More
Cayman Chemical 1MyrIstoyld72LInoleoylsn 100ug
SDP
Catalog No. 502910197
Encompass_Preferred
An internal standard for the quantification of 1-myristoyl-2-linoleoyl-sn-glycero-3-PC by GC- or LC-MS
Catalog No. 50-291-0197 Supplier Cayman Chemical Supplier No. 35675100 UG
An internal standard for the quantification of 1-myristoyl-2-linoleoyl-sn-glycero-3-PC by GC- or LC-MS