Learn More
Cayman Chemical 1-Myristyl-2-hydroxy-sn-glycero-3-PA, Purity: ≥98%, 10 mg
SDP
Catalog No. 502895621
Encompass_Preferred
An LPA derivative and agonist of LPA1 and LPA2; increases calcium mobilization in a reporter assay using HEL 92.1.7 cells (EC50 = 37 nM)
Catalog No. 50-289-5621 Supplier Cayman Chemical Supplier No. 900140810 MG
An LPA derivative and agonist of LPA1 and LPA2; increases calcium mobilization in a reporter assay using HEL 92.1.7 cells (EC50 = 37 nM)