Learn More
Boston Bioproducts Inc Citric Acid (1 N) - 500ml
Catalog No. NC1519896
Encompass
As an acidic chemical reagent, Citric Acid Solution assumes critical roles in many laboratory applications. By controling the concentration of hydrogen ions, Citric Acid enables precise pH adjustments in target solutions, maintaining optimal pH levels in biological systems and experimental environments. It also facilitates the isolation and purification of biomolecules such as proteins, DNA, and RNA, and influences the solubility, charge, and stability of affected solutions.
Catalog No. NC1519896 Supplier Boston Bioproducts Inc Supplier No. BB2702500ML
As an acidic chemical reagent, Citric Acid Solution assumes critical roles in many laboratory applications. By controling the concentration of hydrogen ions, Citric Acid enables precise pH adjustments in target solutions, maintaining optimal pH levels in biological systems and experimental environments. It also facilitates the isolation and purification of biomolecules such as proteins, DNA, and RNA, and influences the solubility, charge, and stability of affected solutions.