Learn More
Cayman Chemical 1Naphthyl 3 5DNItrobenzAT 10mg
SDP
Catalog No. 502952314
Encompass_Preferred
A dual 5-LO and mPGES-1 inhibitor (IC50s = 1 and 3.6 µM, respectively)
Catalog No. 50-295-2314 Supplier Cayman Chemical Supplier No. 1172910 MG
A dual 5-LO and mPGES-1 inhibitor (IC50s = 1 and 3.6 µM, respectively)