Learn More
Enzo Life Sciences 1-O-Hexadecyl-2-O-acetyl-sn-glycerol (5mg). CAS: 77133-35-8
SDP
Catalog No. 502010570
Encompass_Preferred
Inhibits protein kinase C activation by diacylglycerols. Stimulates HL-60 cell differentiation. Biosynthetic precursor of PAF via remodeling and retroconversion/de novo routes. Induces delayed aggregation of rabbit platelets. Mitogenic for smooth muscle cells. Purity: ≥95% (TLC). Solubility: Soluble in 100% ethanol or acetone. Long Term Storage: -20°C.
Catalog No. 50-201-0570 Supplier Enzo Life Sciences Supplier No. BMLEI1500005
Inhibits protein kinase C activation by diacylglycerols. Stimulates HL-60 cell differentiation. Biosynthetic precursor of PAF via remodeling and retroconversion/de novo routes. Induces delayed aggregation of rabbit platelets. Mitogenic for smooth muscle cells. Purity: ≥95% (TLC). Solubility: Soluble in 100% ethanol or acetone. Long Term Storage: -20°C.