Learn More
Cayman Chemical 1OHexadecylsnglycerol 100mg
SDP
Catalog No. 502936618
Encompass_Preferred
An alkyl glyceryl ether; reduces UVB-induced cell death and production of ROS and PGE2 in NHEKs; increases coronary flow and left ventricular developed pressure and reduces MDA formation ex vivo in a rat heart model of ischemia/reperfusion injury
Catalog No. 50-293-6618 Supplier Cayman Chemical Supplier No. 25723100 MG
An alkyl glyceryl ether; reduces UVB-induced cell death and production of ROS and PGE2 in NHEKs; increases coronary flow and left ventricular developed pressure and reduces MDA formation ex vivo in a rat heart model of ischemia/reperfusion injury