Learn More
Cayman Chemical 1-O-Hexadecyl-sn-glycerol, Purity: ≥95%, CAS: 506-03-6, 500 mg
SDP
Catalog No. 502936353
Encompass_Preferred
An alkyl glyceryl ether; reduces UVB-induced cell death and production of ROS and PGE2 in NHEKs; increases coronary flow and left ventricular developed pressure and reduces MDA formation ex vivo in a rat heart model of ischemia/reperfusion injury
Catalog No. 50-293-6353 Supplier Cayman Chemical Supplier No. 25723500 MG
An alkyl glyceryl ether; reduces UVB-induced cell death and production of ROS and PGE2 in NHEKs; increases coronary flow and left ventricular developed pressure and reduces MDA formation ex vivo in a rat heart model of ischemia/reperfusion injury