Learn More
Enzo Life Sciences 1-Oleoyl-2-acetyl-sn-glycerol (10mg). CAS: 86390-77-4
SDP
Catalog No. 502010511
Encompass_Preferred
In vitro potent activator of protein kinase C (PKC). Stimulator of secretion and aggregation in blood platelets. Stimulates formation of actin nucleation sites. Stimulation of pp60c-src phosphorylation in EBV transformed B-cells, 470μM. Alternative name: OAG. Purity: ≥98% (TLC). Solubility: Soluble in DMSO or ethanol. Long Term Storage: -20°C.
Catalog No. 50-201-0511 Supplier Enzo Life Sciences Supplier No. BMLDG1250010
In vitro potent activator of protein kinase C (PKC). Stimulator of secretion and aggregation in blood platelets. Stimulates formation of actin nucleation sites. Stimulation of pp60c-src phosphorylation in EBV transformed B-cells, 470μM. Alternative name: OAG. Purity: ≥98% (TLC). Solubility: Soluble in DMSO or ethanol. Long Term Storage: -20°C.